missing translation for 'onlineSavingsMsg'
Learn More

Albendazole sulfone

Catalog No. AAH2591703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2591703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2591703 Supplier Thermo Scientific Chemicals Supplier No. H2591703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75184-71-3 | C12H15N3O4S | 297.33 g/mol

Albendazole are currently used for chemotherapeutic treatment of AE. Albendazole sulfone is used as scolicidal agents on hydatid cysts(in vitro).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Albendazole are currently used for chemotherapeutic treatment of AE. Albendazole sulfone is used as scolicidal agents on hydatid cysts(in vitro).

Solubility
Soluble in DMSO, and methanol.

Notes
Stor

Chemical Identifiers

CAS 75184-71-3
Molecular Formula C12H15N3O4S
Molecular Weight (g/mol) 297.33
MDL Number MFCD00600775
InChI Key CLSJYOLYMZNKJB-UHFFFAOYSA-N
Synonym albendazole sulfone, unii-1uic88380g, methyl 5-propylsulfonyl 2-benzimidazolecarbamate, methyl 5-propylsulfonyl-1h-benzimidazol-2-yl carbamate, methyl-5-propylsulfonyl-1h-benzimidazol-2-yl carbamate, 5-propylsulfonyl-2-benzimidazolecarbamic acid methyl ester, methyl 6-propylsulfonyl-1h-benzo d imidazol-2-yl carbamate, methyl n-5-propane-1-sulfonyl-1h-1,3-benzodiazol-2-yl carbamate, 2-benzimidazolecarbamic acid, 5-propylsulfonyl-, methyl ester
PubChem CID 53174
ChEBI CHEBI:80620
IUPAC Name methyl N-[6-(propane-1-sulfonyl)-1H-1,3-benzodiazol-2-yl]carbamate
SMILES CCCS(=O)(=O)C1=CC=C2N=C(NC(=O)OC)NC2=C1

Specifications

Melting Point 290°C to 292°C
Quantity 1 g
Solubility Information Soluble in DMSO,and methanol.
Formula Weight 297.33
Chemical Name or Material Albendazole sulfone
Hazard Category H361
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
RTECSNumber DD6520700
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only