missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-Alanine, 98.5%, Thermo Scientific Chemicals

Catalog No. AC163795000
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163795000 500 g Plastic Bottle
AC163790025 2.5 kg Plastic Bottle
AC163790100 10 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC163795000 Supplier Thermo Scientific Chemicals Supplier No. 163795000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 107-95-9 | C3H7NO2 | 89.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-95-9
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00008200
InChI Key UCMIRNVEIXFBKS-UHFFFAOYSA-N
Synonym beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino
PubChem CID 239
ChEBI CHEBI:16958
IUPAC Name 3-aminopropanoic acid
SMILES NCCC(O)=O

Specifications

Melting Point 197.0°C to 202.0°C
Ammonium 1000ppm max.
CAS Min % 98.5
CAS Max % 100.0
pH 6.5 to 7.5 (10% w/v aq. soln.)
Color White
Chlorine (Cl) 400ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Loss on Drying 0.5% max. (105°C, 3 hrs)
Assay Percent Range 99%
Other Amino Acids (TLC) Passes Test (not detectable on 10μgspotting)
Packaging Plastic Bottle
Linear Formula H2NCH2CH2CO2H
Fieser 01,16; 05,6; 06,10; 17,3
Sulfate 480ppm max.
Sulfated Ash 0.2% max.
Quantity 500 g
Formula Weight 89.09
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material β-Alanine
Show More Show Less

RUO – Research Use Only