missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals beta-Alanine, 98.5%, Thermo Scientific Chemicals
CAS: 107-95-9 | C3H7NO2 | 89.09 g/mol
Supplier: Thermo Scientific Chemicals 163790025
| Quantity | 2.5 kg |
|---|---|
| Packaging | Plastic Bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C3H7NO2 | |
| MFCD00008200 | |
| beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino | |
| CHEBI:16958 | |
| NCCC(O)=O |
Specifications
| 197.0°C to 202.0°C | |
| 107-95-9 | |
| 98.5 | |
| 6.5 to 7.5 (10% w/v aq. soln.) | |
| 400ppm max. | |
| Authentic | |
| 0.5% max. (105°C, 3 hrs) | |
| (TLC) Passes Test (not detectable on 10μgspotting) | |
| C3H7NO2 | |
| MFCD00008200 | |
| 480ppm max. | |
| 0.2% max. | |
| NCCC(O)=O | |
| 2.5 kg | |
| 239 | |
| 89.09 | |
| Crystalline Powder |
| β-Alanine | |
| 1000ppm max. | |
| 100.0 | |
| White | |
| 10ppm max. | |
| 30ppm max. | |
| 99% | |
| Plastic Bottle | |
| H2NCH2CH2CO2H | |
| 01,16; 05,6; 06,10; 17,3 | |
| beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino | |
| UCMIRNVEIXFBKS-UHFFFAOYSA-N | |
| 3-aminopropanoic acid | |
| 89.09 | |
| CHEBI:16958 | |
| 99% |