ACROBiosystems Inc Recombinant Protein;Human Akt1 Protein, His Tag & Strep Tag;HEK293;500UG;AK1,AKT1,PKB,RAC,RAC-PK-alpha,PKB alpha,AKT-1
Catalog No. 501659068
Recombinant Protein;Human Akt1 Protein, His Tag & Strep Tag;500UG;Human Akt1, His Tag, Strep Tag (AK1-H5283) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 480 (Accession # AAH00479).;This protein carries a polyhistidine tag at the C-terminus, followed by a twin strep tag. The protein has a calculated MW of 59.5 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Human Akt1 Protein, His Tag & Strep Tag;500UG;Human Akt1, His Tag, Strep Tag (AK1-H5283) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 480 (Accession # AAH00479).;This protein carries a polyhistidine tag at the C-terminus, followed by a twin strep tag. The protein has a calculated MW of 59.5 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.