Learn More
AirClean™ Systems HEPA and ULPA Filters
For removal of powders, fibers and particulate
Supplier:  AirClean™ Systems ACFULPA
 Afficher plus de versions de ce produit
Numéro de catalogue. 361018804
Il en reste null
Description
Description
  • Electrostatically charged filters prevent “particle-rain“
  • Metal screened on face of filter to promote even air distribution and laminar airflow
  • Metal-free filters available upon request
Spécifications
Identifiants chimiques
Air Filter
Spécifications
99.9995%
ACFULPA
Air Filter
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS