missing translation for 'onlineSavingsMsg'
Learn More

AirClean™ Systems Combination PCR Workstations

Numéro de catalogue. 360993862
Change view
Click to view available options
Dimensions (D x W x H):
24 x 32 x 30 in.
24 x 32 x 41 in.
24 x 48 x 30 in.
24 x 48 x 41 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Dimensions (D x W x H)
360993862 24 x 48 x 30 in.
361004288 24 x 32 x 41 in.
361004289 24 x 48 x 41 in.
360993859 24 x 32 x 30 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 360993862 Fournisseur AirClean™ Systems Code fournisseur AC648LFUVC

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

The Airclean Combination PCR Workstation combines ISO 5 (FS209E) Class 100 clean air environment with UV light sterilization for optimal protection from sample contamination.

  • UVTect™ microprocessor controller
  • Class 100 (ISO 5) vertical laminar flow air
  • Polycarbonate and polypropylene design to reflect UV energy
  • Full access folding sash
  • Unibody plastic design - no joints or gaps in construction
  • Digital UV light timer 0-59 minutes
  • Lab event timer
  • Available in 24 in. , 32 in., and 48 in. widths; nominal depth 24 in.
  • 110V AC or 220V AC versions (24 in. only offered as 110V)

UVTect™ Features:

  • Variable speed blower control
  • 254nm ultraviolet light with user set digital timer
  • Constant HEPA filter monitoring
  • Constantly monitors and alerts effectiveness of the shortwave UV bulb life
  • Lab event timer

Spécifications

Product Type Combination PCR Workstation
Lighting Type Digital UV
Dimensions (D x W x H) 24 x 48 x 30 in.