missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ AIN-93M Rodent Diet

Catalog No. 14726618
Encompass
Change view
Click to view available options
Format:
Powder
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Quantity
14726618 Powder 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14726618 Supplier Bio-Serv™ Supplier No. F3198

Purified diet formulated to meet the nutritional requirements of mature rodents

  • Widely used and published
  • Phytoestrogen free
  • Chlorophyll free—does not interfere with imaging studies
  • Nutritionally assayed
  • Ingredients: Corn Starch, Casein, Maltodextrin, Sucrose, Cellulose, Soybean Oil, Mineral Mix, Vitamin Mix, Choline Bitartrate, L-Cystine, tBHQ

Specifications

Product Type Rodent Diet
Components Corn Starch, Casein, Maltodextrin, Sucrose, Soybean Oil, Cellulose, Mineral Mix, Vitamin Mix, L-Cystine, Choline Bitartrate, tBHQ
Format Powder
For Use With (Application) Used and published for research objectives
Particle Shape Powder
Quantity 1 kg
Species Rodent