Learn More
Across International Llc Ai 200C 15L SST Compact Desktop Heated Recirculator 220V
Numéro de catalogue. 502251316
Encompass
The Ai H200 stainless steel series heated recirculators with 2,800 watts heater offer outstanding performance and high reliability as well as quiet operation and ease-of-use. Powerful force/suction pump provides constant flow rates, while the 2nd generation PID controller gives you the ideal solution when you need precise temperature control.
Numéro de catalogue. 502251316 Fournisseur Across International Llc Code fournisseur H20015SST.220
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The Ai H200 stainless steel series heated recirculators with 2,800 watts heater offer outstanding performance and high reliability as well as quiet operation and ease-of-use. Powerful force/suction pump provides constant flow rates, while the 2nd generation PID controller gives you the ideal solution when you need precise temperature control.