missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AGTR1 Recombinant Rabbit Monoclonal Antibody (23GB635)

Numéro de catalogue. MA553169
Modifier l'affichage
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MA553169 20 μL
MA553168 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MA553169 Fournisseur Invitrogen™ Code fournisseur MA553169
Il en reste null

Rabbit Recombinant Monoclonal Antibody

AGTR1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

AGTR1 is known as Angiotensin II Type 1 Receptor. Angiotensin II is a potent vasopressor hormone and a primary regulator of aldosterone secretion. It is an important effector controlling blood pressure and volume in the cardiovascular system. And it acts through at least two types of receptors. This gene encodes the type 1 receptor which is thought to mediate the major cardiovascular effects of angiotensin II. Moreover, this gene may play a role in the generation of reperfusion arrhythmias following restoration of blood flow to ischemic or infarcted myocardium. It was previously thought that a related gene, denoted as AGTR1B, existed; however, it is now believed that there is only one type 1 receptor gene in humans. Multiple alternatively spliced transcript variants have been reported for this gene.

Spécifications

Antigen AGTR1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB635
Concentration 2.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AGTR1
Gene Accession No. P30556
Gene Alias 1810074K20Rik; AG2S; AGTR1; Agtr1a; Agtr-1a; AGTR1B; AI551199; angiotensin II receptor type 1; angiotensin II receptor, type 1; angiotensin II receptor, type 1a; Angiotensin II type-1 receptor; Angiotensin II type-1A receptor; angiotensin receptor 1; angiotensin receptor 1a; Angtr-1a; AT1; At1a; AT1AR; AT1B; AT1BR; AT1R; AT2R1; AT2R1A; AT2R1B; HAT1R; Type-1 angiotensin II receptor; Type-1A angiotensin II receptor; type-1B angiotensin II receptor; vascular type-1 angiotensin II receptor
Gene Symbols AGTR1, Agtr1a
Host Species Rabbit
Immunogen A synthetic peptide of human AGTR1 (1-48AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11607, 185
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats