Learn More
Brady™ Aggressive Adhesive Multipurpose Nylon Labels
The B-499 nylon cloth label is made of tough and versatile, general-purpose label material. Print almost anywhere with a Brady™ portable label printer.
Supplier:  Brady™ M411499
 Afficher plus de versions de ce produit
Numéro de catalogue. 17012225
Il en reste null
Description
Description
  • Ideal for labeling tightly-curved surfaces such as wires and cables, as well as general-purpose lab, equipment panels, faceplates and terminal blocks
  • Designed to withstand extreme temperatures, chemicals, greases, oils, cleaners and detergents
  • Permanent adhesive labels that stick to smooth, rough and powder-coated surfaces alike
Spécifications
Identifiants chimiques
UL Recognised;CSA Accepted;Halogen Free (DIN VDE 0472/part 815) Approvals are for White material only Compliant with Directive 2011/65/EU and 2015/863
White
Nylon Cloth
-40°F to 194°F
0.0065 in
Permanent Acrylic
Matte
Non-sterile
-40°C to 90°C
0.166 mm
Spécifications
UL Recognised;CSA Accepted;Halogen Free (DIN VDE 0472/part 815) Approvals are for White material only Compliant with Directive 2011/65/EU and 2015/863
White
Die-cut
Label
Component and equipment labelling;Electrical labelling;General identification;Warehouse marking;Wire and cable labelling
220
-40°C to 90°C
0.166 mm
0.5 in.
19.05 mm
12.70 mm
Permanent Acrylic
Matte
Nylon Cloth
Non-sterile
BMP41;BMP51;BMP53;M511;M410;M510
-40°F to 194°F
0.0065 in
19.05 mm (W) x 12.70 mm (H)
0.75 in
0.5 in
M4-11-499
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS