missing translation for 'onlineSavingsMsg'
Learn More

Agarose LE, Molecular Biology Grade, Ultrapure

CAS: 9012-36-6

$152.45 - $2,870.55

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AAJ3280214
View Documents Promotion Details
N/A 25 g Plastic Bottle
Each for $152.45
Only null left
 
AAJ3280222
View Documents Promotion Details
N/A 100 g Plastic Bottle
Each for $424.81
Only null left
 
AAJ3280230
View Documents Promotion Details
N/A 250 g Plastic Bottle
Each for $893.33
Only null left
 
AAJ3280236
View Documents Promotion Details
N/A 500 g Plastic Bottle
Each for $1,618.84
Only null left
 
AAJ32802A1
View Documents Promotion Details
N/A 1 kg Plastic Bottle
Each for $2,870.55
Only null left
 
Description

Description

Specifications

Chemical Identifiers

9012-36-6
MFCD00081294

Specifications

Granular Free-flowing Powder
MFCD00081294
Agarose LE
9012-36-6
Molecular Biology
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-731-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only