missing translation for 'onlineSavingsMsg'
Learn More

Agarose LE, Molecular Biology Grade, Ultrapure

CAS: 9012-36-6

Supplier:  Thermo Fisher Scientific J3280230

Quantity 250 g
Packaging Plastic Bottle

 View more versions of this product

Catalog No. AAJ3280230


Only null left

Description

Description

Specifications

Chemical Identifiers

9012-36-6
MFCD00081294

Specifications

Agarose LE
9012-36-6
Plastic Bottle
Molecular Biology
Granular Free-flowing Powder
250 g
MFCD00081294
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-731-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions