missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agarose LE, Molecular Biology Grade, Ultrapure
CAS: 9012-36-6
Supplier: Thermo Fisher Scientific J3280222
| Quantity | 100 g |
|---|---|
| Packaging | Plastic Bottle |
Chemical Identifiers
| 9012-36-6 | |
| MFCD00081294 |
Specifications
| Agarose LE | |
| 9012-36-6 | |
| Plastic Bottle | |
| Molecular Biology |
| Granular Free-flowing Powder | |
| 100 g | |
| MFCD00081294 |
Safety and Handling
EINECSNumber : 232-731-8
TSCA : Yes
Recommended Storage : Ambient temperatures