missing translation for 'onlineSavingsMsg'
Learn More

Agarose LE, Molecular Biology Grade, Ultrapure

Catalog No. AAJ3280214
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
Packaging:
Plastic Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ3280214 25 g Plastic Bottle
AAJ3280222 100 g Plastic Bottle
AAJ3280230 250 g Plastic Bottle
AAJ3280236 500 g Plastic Bottle
AAJ32802A1 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AAJ3280214 Supplier Thermo Fisher Scientific Supplier No. J3280214

CAS: 9012-36-6

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Granular Free-flowing Powder
Quantity 25 g
Packaging Plastic Bottle
Grade Molecular Biology
Chemical Name or Material Agarose LE
EINECSNumber 232-731-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only