missing translation for 'onlineSavingsMsg'
Learn More

Agar, Powder, FCC, Spectrum™ Chemical

Catalog No. 18600331
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
25 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600331 125 g Poly Bottle
18600332 500 g Poly Bottle
18600333 2.5 kg Poly Bottle
18600334 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600331 Supplier Spectrum Chemical Mfg Cor Supplier No. A1672125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

A1672, 9002-18-0

Spectrum™ Chemical Agar, Powder, FCC is a gel made from the cell walls of several species of red algae or seaweed. It appears as an odorless, tasteless, beige powder. It functions as a binder and thickener in cooking to make jellies, pudding, and custards. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9002-18-0
Molecular Formula C14H24O9
Molecular Weight (g/mol) 336.34
InChI Key GYYDPBCUIJTIBM-UHFFFAOYNA-N
IUPAC Name 2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
SMILES COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Color Beige to Brown
Quantity 125 g
Grade FCC
Chemical Name or Material Agar