Learn More
Eppendorf™ Aerosol-Tight Cap, Universal Buckets, S-4xUniversal Rotor
Catalog No. 13864444
Change view
Click to view available options
For Use With (Equipment):
S-4xUniversal rotor buckets
Catalog No. For Use With (Equipment)
13864444 S-4xUniversal rotor buckets
1 options
Catalog No. 13864444 Supplier Eppendorf™ Supplier No. 5910750005
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Seals centrifuge rotor buckets for protection against hazardous materials.
  • Suitable for S-4xUniversal Rotor buckets
  • Creates aerosol-tight seal when working with hazardous substances
Specifications
For Use With (Equipment) S-4xUniversal rotor buckets