Learn More
OHAUS Adventurer Semi-Micro Balance
Striking the ideal balance between inventive features and functional, uncomplicated weighing capabilities, the OHAUS Adventurer incorporates all of the applications necessary for routine weighing and measurement activities.
Supplier: OHAUS 30852314
Description
Adventurer balances feature a color touchscreen, icon-based user interface, and an ergonomic design, making them easy to configure and use.
- Features such as specialized weighing modes, multiple connectivity options, and AutoCal™ provide versatility and flexibility for a variety of applications.
- Durable construction, large weighing surfaces, a space-saving draftshield design, and full housing in-use cover allow for use in lab, education and industrial environments.
Specifications
| 82 g, 120 g | |
| 0.00001 g | |
| Balance |
| LCD | |
| AX85 | |
| No |