missing translation for 'onlineSavingsMsg'
Learn More

Advantame™, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015331
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015331 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015331 Supplier Merck Emd Millipore Supplier No. 80054100MG

Advantame(R) is a high intensity, non-caloric sweetener, which is derived from aspartame. It is widely used in food and beverage industry.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line Advantame
CAS 714229-20-6
Linear Formula C24H30N2O7 · H2O
Synonym N-[N-[3-(3-Hydroxy-4-methoxyphenyl)propyl]-alpha-L-aspartyl]-L-phenylalanine 1-methyl ester monohydrate
Type Analytical Standard
Quantity 100 mg
Formula Weight 476.52
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard

Advantame is a registered trademark of Ajinomoto Co., Inc.