Learn More
Advanced Instruments Sampler Calibration Key for Advanced™ 3320 Micro Osmometer
Catalog No. 22046741
Change view
Click to view available options
For Use With (Equipment):
3320 Micro-osmometer
Catalog No. For Use With (Equipment)
22046741 3320 Micro-osmometer
1 options
Catalog No. 22046741 Supplier Advanced Instruments Supplier No. 3M0827
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Replacement calibration key for Advanced 3320 Micro Osmometer 20μL sampler
Specifications
For Use With (Equipment) 3320 Micro-osmometer
Type Calibration Gauge
Intended Use Serum, Urine or other bodily fluids in clinical applications and media or formulations in pharmaceutical, biotech or industrial applications
Quantity Each