missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adonitol, 99%

CAS: 488-81-3 | C5H12O5 | 152.146 g/mol

Supplier:  Thermo Scientific Chemicals L0325314

 View more versions of this product

Catalog No. AAL0325314


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Adonitol is metabolized to teicholic acids used in the cell walls of gram positive bacteria. Adonitol is often compared to other cell permeating molecules such as formamide, propanediol, and DMSO as a cryopreservation agent.

Solubility
Soluble in water (100 mg/ml).

Notes
Hygroscopic. Store under dry inert gas. Incompatible with water and oxidizing agents.
Specifications

Specifications

Adonitol
102°C to 104°C
MFCD00064291
1720524
14,168
Soluble in water (100mg/ml).
C(C(C(C(CO)O)O)O)O
152.146
152.15
488-81-3
C5H12O5
25 g
Hygroscopic
xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
HEBKCHPVOIAQTA-NGQZWQHPSA-N
(2S,4R)-pentane-1,2,3,4,5-pentol
6912
99%

Chemical Identifiers

488-81-3
152.146
HEBKCHPVOIAQTA-NGQZWQHPSA-N
6912
C(C(C(C(CO)O)O)O)O
C5H12O5
MFCD00064291
xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
(2S,4R)-pentane-1,2,3,4,5-pentol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-685-7

RTECSNumber : VJ0800000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only