accessibility menu, dialog, popup

UserName

HOOHOHOHOH

CAS RN 488-81-3

IUPAC Name: pentane-1,2,3,4,5-pentol
Synonyms: 1,2,3,4,5-pentahydroxypentane
Molecular Weight (g/mol): 152.15
Molecular Formula: C5H12O5
InChi Key: HEBKCHPVOIAQTA-UHFFFAOYNA-N
SMILES: OCC(O)C(O)C(O)CO
Molecular Weight (g/mol) 
  • (7)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (4)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (6)
  • (7)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)