missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adonitol, 99%

CAS: 488-81-3 | C5H12O5 | 152.146 g/mol

$59.19 - $219.36

Chemical Identifiers

CAS 488-81-3
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.146
MDL Number MFCD00064291
InChI Key HEBKCHPVOIAQTA-NGQZWQHPSA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
SMILES C(C(C(C(CO)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAL0325306
View Documents Promotion Details
Thermo Scientific Chemicals
L0325306
5 g
Each for $59.19
Only null left
 
AAL0325314
View Documents Promotion Details
Thermo Scientific Chemicals
L0325314
25 g
Each for $219.36
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Adonitol is metabolized to teicholic acids used in the cell walls of gram positive bacteria. Adonitol is often compared to other cell permeating molecules such as formamide, propanediol, and DMSO as a cryopreservation agent.

Solubility
Soluble in water (100 mg/ml).

Notes
Hygroscopic. Store under dry inert gas. Incompatible with water and oxidizing agents.
Specifications

Chemical Identifiers

488-81-3
152.146
HEBKCHPVOIAQTA-NGQZWQHPSA-N
6912
C(C(C(C(CO)O)O)O)O
C5H12O5
MFCD00064291
xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
(2S,4R)-pentane-1,2,3,4,5-pentol

Specifications

488-81-3
C5H12O5
1720524
14,168
Soluble in water (100mg/ml).
C(C(C(C(CO)O)O)O)O
152.146
152.15
Adonitol
102°C to 104°C
MFCD00064291
Hygroscopic
xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
HEBKCHPVOIAQTA-NGQZWQHPSA-N
(2S,4R)-pentane-1,2,3,4,5-pentol
6912
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-685-7

RTECSNumber : VJ0800000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only