missing translation for 'onlineSavingsMsg'
Learn More

Adipoyl Chloride 98.0+%, TCI America™

Catalog No. A0169500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0169500G 500 g
A016925G 25 g
A0169100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0169500G Supplier TCI America Supplier No. A0169500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-50-2
Molecular Formula C6H8Cl2O2
Molecular Weight (g/mol) 183.028
MDL Number MFCD00000759
InChI Key PWAXUOGZOSVGBO-UHFFFAOYSA-N
Synonym adipoyl chloride, adipoyl dichloride, adipyl chloride, adipoylchloride, adipic dichloride, hexanedioyl chloride, adipic acid dichloride, butane-1,4-dicarbonyl chloride, adipic acid chloride, pubchem13242
PubChem CID 61034
IUPAC Name hexanedioyl dichloride
SMILES C(CCC(=O)Cl)CC(=O)Cl

Specifications

Color Yellow
Boiling Point 115°C
Quantity 500 g
UN Number 3265
Formula Weight 183.03
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Adipoyl Chloride
EINECSNumber (2)-2563
TSCA Yes