missing translation for 'onlineSavingsMsg'
Learn More

Adipoyl chloride, 98%

Catalog No. AAA1316836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1316836 500 g
AAA1316814 25 g
AAA1316822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1316836 Supplier Thermo Scientific Chemicals Supplier No. A1316836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-50-2 | C6H8Cl2O2 | 183.028 g/mol

Adipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.

Solubility
Immiscible with water.

Notes
Incompatible strong oxidizing agents, bases and acids.

Chemical Identifiers

CAS 111-50-2
Molecular Formula C6H8Cl2O2
Molecular Weight (g/mol) 183.028
MDL Number MFCD00000759
InChI Key PWAXUOGZOSVGBO-UHFFFAOYSA-N
Synonym adipoyl chloride, adipoyl dichloride, adipyl chloride, adipoylchloride, adipic dichloride, hexanedioyl chloride, adipic acid dichloride, butane-1,4-dicarbonyl chloride, adipic acid chloride, pubchem13242
PubChem CID 61034
IUPAC Name hexanedioyl dichloride
SMILES C(CCC(=O)Cl)CC(=O)Cl

Specifications

Density 1.255
Boiling Point 105°C to 107°C (2 mmHg)
Flash Point >110°C (230°F)
Odor Acrid
Refractive Index 1.4705
Linear Formula ClCO(CH2)4COCl
Quantity 500 g
UN Number UN3265
Beilstein 507709
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 183.03
Percent Purity 98%
Chemical Name or Material Adipoyl chloride
Show More Show Less
Hazard Category H227-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 203-876-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only