Learn More
Electron Microscopy Sciences SuperFrost™ PLUS Adhesion Slide, Blue
Numéro de catalogue. 50949531
Encompass
Superfrost Plus glass slides feature:
They place a permanent positive charge in the microscope slide.
They electrostatically attract frozen tissue sections and cytology preparations, binding them to the slide.
They form a bridge so that covalent bonds develop between formalin fixed sections and the glass.
Tissue sections and cytological preparations adhere better to the Plus glass slides without the need for special adhesives or protein coatings.
Plus glass puts an end to background staining and eliminates blue or red background staining which is often encountered in hematoxylin and eosin staining and brown background staining in immuno-peroxidase or in situ DNA proceduress.
Plus allow for a higher degree of enzyme digestion without the loss of tissue sections and cytological preparations placed on Plus glass resist cell loss. Plus glass improves tissue adherence in microwave staining.
Color: Blue
Numéro de catalogue. 50949531 Fournisseur Electron Microscopy Sciences Code fournisseur 7186920
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Superfrost Plus glass slides feature:
They place a permanent positive charge in the microscope slide.
They electrostatically attract frozen tissue sections and cytology preparations, binding them to the slide.
They form a bridge so that covalent bonds develop between formalin fixed sections and the glass.
Tissue sections and cytological preparations adhere better to the Plus glass slides without the need for special adhesives or protein coatings.
Plus glass puts an end to background staining and eliminates blue or red background staining which is often encountered in hematoxylin and eosin staining and brown background staining in immuno-peroxidase or in situ DNA proceduress.
Plus allow for a higher degree of enzyme digestion without the loss of tissue sections and cytological preparations placed on Plus glass resist cell loss. Plus glass improves tissue adherence in microwave staining.
Color: Blue