missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADFP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580906
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PLIN2.
Milk lipid globules from humans, cows and rats contain a protein identified as adipocyte differentiation-related protein (ADFP). It is associated with the globule surface membrane material. This protein, previously believed to be specific to adipocytes, is a major constituent of the globule surface and is present in a detergent-insoluble complex that contains butyrophilin and xanthine oxidase. ADFP (Adipophilin) is found in a wide range of cultured cell lines, including fibroblasts, endothelial and epithelial cells. In tissues, however, expression of adipophilin is restricted to specific cell types, such as lactating mammary epithelial cells, adrenal cortex cells, Sertoli and Leydig cells of the male reproductive system, and steatosis or fatty change hepatocytes in alcoholic liver cirrhosis. ADFP may be a possible new marker for the identification of specialized differentiated cells containing lipid droplets and for diseases associated with fat-accumulating cells.Identifiants chimiques
| ADFP | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| Plin2 | |
| AA407157; Adfp; Adipophilin; adipose differentiation related protein; adipose differentiation-related protein; ADPH; Adrp; ADRP antibody; MGC10598; perilipin 2; perilipin-2; Plin2 | |
| Plin2 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with no preservative | |
| Q99541 | |
| 123 | |
| A synthetic peptide corresponding to the N-terminus of the Human PLIN2. | |
| Antibody |
Specifications
| ADFP | |
| Polyclonal | |
| Unconjugated | |
| Plin2 | |
| AA407157; Adfp; Adipophilin; adipose differentiation related protein; adipose differentiation-related protein; ADPH; Adrp; ADRP antibody; MGC10598; perilipin 2; perilipin-2; Plin2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 123 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q99541 | |
| Plin2 | |
| A synthetic peptide corresponding to the N-terminus of the Human PLIN2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |