Learn More
Thermo Scientific Chemicals Adenosine, 99%
CAS: 58-61-7 | C10H13N5O4 | 267.25 g/mol
Supplier: Thermo Scientific Chemicals A1078118
| Quantity | 50 g |
|---|
Description
Adenosine serves as a nucleotide. It also plays an important role in the regulation of blood flow to various organs through vasodilation. It is actively involved in biochemical processes, such as energy transfer as adenosine triphosphate and adenosine diphosphate. Further, it is used in medication specifically as an antiarrhythmic agent to treat a number of supraventricular tachycardia. It is used to promote thickening of hair for people with thinning hair.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water, ammonium hydroxide and dimethyl sulfoxide. Insoluble in ethanol.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Identifiants chimiques
| 58-61-7 | |
| 267.25 | |
| OIRDTQYFTABQOQ-KQYNXXCUSA-N | |
| 60961 | |
| (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
| C10H13N5O4 | |
| MFCD00005752 | |
| adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol | |
| CHEBI:16335 | |
| NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1 |
Spécifications
| Adenosine | |
| 234°C to 237°C | |
| C10H13N5O4 | |
| MFCD00005752 | |
| 14,153 | |
| Soluble in water,ammonium hydroxide and dimethyl sulfoxide. Insoluble in ethanol. | |
| NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1 | |
| 267.25 | |
| CHEBI:16335 | |
| 99% |
| 58-61-7 | |
| 99% | |
| 50 g | |
| 93029 | |
| adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol | |
| OIRDTQYFTABQOQ-KQYNXXCUSA-N | |
| (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol | |
| 60961 | |
| 267.25 |
Sécurité et manipulation
EINECSNumber : 200-389-9
RTECSNumber : AU7175000
TSCA : Yes
Recommended Storage : Keep cold