missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adénosine, 99+%

Catalog No. AC164040050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Bouteille en verre
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164040050 5 g Bouteille en verre
AC164040250 25 g Bouteille en verre
AC164041000 100 g Bouteille en verre
AC164045000 500 g Bouteille en verre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC164040050 Supplier Thermo Scientific Chemicals Supplier No. 164040050
Obtenez jusqu’à 1,800 $ en produits chimiques
View all promotions

CAS : 58-61-7 | C10H13N5O4 | 267,25 g/mol

Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58-61-7
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00005752
InChI Key OIRDTQYFTABQOQ-KQYNXXCUSA-N
Synonym adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol
PubChem CID 60961
ChEBI CHEBI:16335
IUPAC Name (2R,3R,4S,5R)-2-(6-aminopurine-9-yl)-5-(hydroxyméthyl)oxolane-3,4-diol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Absorbance 0.76 to 0.80 (A250/A260),0.14 to 0.16 (A280/A260)
Melting Point 233.0°C to 238.0°C
Color Blanc
Loss on Drying 0.5% (105°C, 2 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Bouteille en verre
Quantity 5 g
Beilstein 26, V,16, 335
Merck Index 15141
Specific Rotation − 60.00
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Spectrophotometry 98.0 to 102% (lambda max. 259nm,pH 7) molar absorptivity 15400+/-2% ,on dry basis
Transmittance 98% (c=0.5, H2O, 430nm) 1cm cell min.
Formula Weight 267.23
Percent Purity 99+%
Physical Form Poudre cristalline
Chemical Name or Material Adenosine
Show More Show Less
EINECSNumber 200-389-9

RUO – Usage réservé à la recherche