missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine-3',5'-cyclic monophosphate sodium salt, 99%

Catalog No. AAJ62174MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62174MD 250 mg
AAJ6217403 1 g
AAJ6217406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ62174MD Supplier Thermo Scientific Chemicals Supplier No. J62174MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 37839-81-9 | C10H11N5NaO6P | 351.19 g/mol

Adenosine 3?,5?-cyclic monophosphate sodium salt monohydrate is used as an activator of protein kinase A (PKA) . A key regulator of many cellular reactions. Released in response to hormonal stimulation. . Directly activates Epac, a Rap1 guanine-nucleotide exchange factor. Acts as a second messenger in signal transduction mechanisms. Promotes apoptosis in resting human B lymphocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate is used as an activator of protein kinase A (PKA) . A key regulator of many cellular reactions. Released in response to hormonal stimulation. . Directly activates Epac, a Rap1 guanine-nucleotide exchange factor. Acts as a second messenger in signal transduction mechanisms. Promotes apoptosis in resting human B lymphocytes.

Solubility
Soluble in water (50 mg/ml).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 37839-81-9
Molecular Formula C10H11N5NaO6P
Molecular Weight (g/mol) 351.19
MDL Number MFCD00069736
InChI Key BXJBFCKTIWRKMQ-GWKNMROSNA-M
Synonym adenosine 3',5'-cyclic monophosphate sodium salt monohydrate, c10h11n5o6p.na.h2o, sodium 3',5'-cyclic amp anion hydrate, adenosine 3',5'-cyclic monophosphate monosodium salt
PubChem CID 91872444
IUPAC Name sodium;(4aR,6R,7R,7aS)-6-(6-aminopurin-9-yl)-2-oxido-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol;hydrate
SMILES [Na+].NC1=C2N=CN([C@@H]3O[C@@H]4COP([O-])(=O)O[C@H]4[C@H]3O)C2=NC=N1

Specifications

Color White
Quantity 250 mg
Beilstein 54612
Solubility Information Soluble in water (50mg/ml).
Formula Weight 351.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Adenosine-3',5'-cyclic monophosphate sodium salt
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only