missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine, 99%

Catalog No. AAA1490636
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1490636 500 g
AAA1490614 25 g
AAA1490618 50 g
AAA1490630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1490636 Supplier Thermo Scientific Chemicals Supplier No. A1490636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 73-24-5 | C5H5N5 | 135.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Solubility
Soluble in water (3 g/L) at 20°C.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 73-24-5
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00041790
InChI Key GFFGJBXGBJISGV-UHFFFAOYSA-N
Synonym adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
PubChem CID 190
ChEBI CHEBI:16708
IUPAC Name 7H-purin-6-amine
SMILES NC1=C2NC=NC2=NC=N1

Specifications

Melting Point >360°C (decomposition)
Odor Odorless
Assay Percent Range 99%
Quantity 500 g
UN Number UN2811
Beilstein 5777
Merck Index 14,152
Solubility Information Soluble in hot water,methanol and ethanol. Slightly soluble in cold water. Insoluble in diethyl ether.
Formula Weight 135.13
Percent Purity 99%
Chemical Name or Material Adenine
Show More Show Less
Hazard Category H301-H500
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-796-1
RTECSNumber AU6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only