missing translation for 'onlineSavingsMsg'
Learn More

Cytiva ADCF-MAb Media

Numéro de catalogue. p-7058049
Change view
Click to view available options
Quantity:
500 mL
1000 mL
6 x 1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
SH3034901 500 mL
SH30349LS 6 x 1000 mL
SH3034902 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. SH3034901 Fournisseur Cytiva Code fournisseur SH30349.01
Il en reste null

Increase process yields for the manufacture of monoclonal antibodies (MAbs) with HyClone™ ADCF-MAb Media, a protein-free media containing no animal-derived components.

  • ADCF
  • Metabolically designed for high cell yield and monoclonal antibody production
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
  • Contains L-glutamine and Poloxamer 188
  • Formulations without L-glutamine support the GS gene expression system

Spécifications

Form Liquid
Product Type ADCF-mAb Liquid Medium
Sterilization Method 0.2 μm Sterile-filtered
With Additives Glutamine, Poloxamer 188, HEPES, Sodium Bicarbonate
Without Additives No Protein, No Phenol Red
Quantity 500 mL
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light
Description with L-Glutamine