Learn More
Promega ADCC Reporter Bioassay, Target (WIL2-S)
Catalog No. PRG7013
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRG7013 1 Ea.
1 options
Catalog No. PRG7013 Supplier Promega Supplier No. G7013
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
  • Control Ab, Anti-CD20
  • ADCC Bioassay Target Cells (WIL2-S)
Recommended for use as a control with all Fc Effector Bioassay Core Kits. ADCC Reporter Bioassay provides a biologically relevant and specific mechanism-of-action-based measure of ADCC without the complex workflow and variability inherent in primary cell-based assays.
Specifications
Quantity 1 Ea.
Content And Storage Varies Among Components