Learn More
Promega ADCC Reporter Bioassay, Complete (WIL2-S)
Catalog No. PRG7014
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRG7014 1 Ea.
1 options
Catalog No. PRG7014 Supplier Promega Supplier No. G7014
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
  • ADCC Bioassay Effector Cells
  • RPMI 1640 Medium
  • Low IgG Serum
  • Bio-Glo™ Luciferase Assay Buffer
  • Bio-Glo™ Luciferase Assay Substrate
  • Control Ab, Anti-CD20
  • ADCC Bioassay Target Cells (WIL2-S)
Provides a biologically relevant and specific mechanism-of-action-based measure of ADCC without the complex workflow and variability inherent in primary cell-based assays. The bioassay also can be used to quantify effects of antibody glycosylation on Fc effector function.
Specifications
Quantity 1 Ea.
Content And Storage Varies Among Components