missing translation for 'onlineSavingsMsg'
Learn More

Promega ADCC Bioassay Effector Cells, F Variant, Propagation Model

Catalog No. PRG9302
Encompass
Change view
Click to view available options
Quantity:
2 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG9302 2 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG9302 Supplier Promega Supplier No. G9302

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

Allows propagation and banking of the ADCC Bioassay, F Variant, Effector Cells without the complex workflow and variability inherent in primary cell-based assays. The bioassay also can be used to quantify effects of antibody glycosylation on Fc effector function.

  • Easily quantitate impact of FcγRIIa polymorphism on Fc effector function
  • Measure antibody potency and stability
  • Includes two vials of Bio-Glo™ Luciferase Assay System reagent at 2x107 cells/mL and 0.65mL/vial, for creating a cell bank and as a backup

Specifications

Quantity 2 x 1 mL
Content And Storage Store below -140°C (freezer or liquid nitrogen vapor phase)
For Use With (Application) Allows propagation and banking of the ADCC Bioassay, F Variant, Effector Cells
Product Type ADCC Bioassay Effector Cells, F Variant (CPM)