missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Adapter

Adapter for 100 mL rectangular bucket in Rotor A-4-44.

Supplier:  Eppendorf™ 022637649

 Afficher plus de versions de ce produit

Numéro de catalogue. 05414136


Il en reste null

Description

Description

  • For round-bottom tubes
  • For 100 mL rectangular bucket in Rotor A-4-44
  • 2 pcs.
Spécifications

Identifiants chimiques

Adapter
Round Bottom
Rotor A-4-44

Spécifications

Adapter
2 Tubes
18 to 30 mL
Round Bottom
Rotor A-4-44
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions