missing translation for 'onlineSavingsMsg'
Learn More

Acycloguanosine, 98%

Catalog No. AC445240100
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC445240100 10 mg Glass bottle
AC445240500 50 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AC445240100 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 445240100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 59277-89-3 | C8H11N5O3 | 225.21 g/mol

Chemical Identifiers

CAS 59277-89-3
Molecular Formula C8H11N5O3
Molecular Weight (g/mol) 225.21
MDL Number MFCD00057880
InChI Key MKUXAQIIEYXACX-UHFFFAOYSA-N
Synonym acyclovir, aciclovir, acycloguanosine, zovirax, vipral, virorax, wellcome-248u, 9-2-hydroxyethoxy methyl guanine, zovir, aciclovirum
PubChem CID 2022
ChEBI CHEBI:2453
IUPAC Name 2-amino-9-(2-hydroxyethoxymethyl)-3H-purin-6-one
SMILES NC1=NC2=C(N=CN2COCCO)C(=O)N1

Specifications

Melting Point 256°C to 260°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 mg
Merck Index 15134
Solubility Information Solubility in water: 0.7mg/mL
Formula Weight 225.2
Percent Purity 98%
Chemical Name or Material Acycloguanosine
missing translation for 'healthHazard1' Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
missing translation for 'healthHazard2' Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation
EINECSNumber 261-685-1

RUO – Research Use Only