missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Activated Calf Thymus DNA

Catalog No. 45001368
Change view
Click to view available options
Quantity:
25 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45001368 25 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45001368 Supplier Cytiva Supplier No. 27457501
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-quality activated calf thymus DNA

  • Treated with DNase I

For preparing substrate used in the study of DNA polymerases

Specifications

Format Liquid
Product Type Activated Calf Thymus DNA
Source Viral and eukaryote
Concentration 25 μg/mL
Quantity 25 Units
For Use With (Application) Study of DNA Polymerase