missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Actinomycin D
Description
- Acts by interfering with mRNA synthesis
- ctinomycin D is used as a selective antibiotic for resistant bacteria, generally at a concentration of about 1μg/mL
- Product is provided as a powder
- Made into a stock solution at 1mg/ml in DMSO
Cell Analysis, Cell Structure, Cloning, Nucleus, Nucleoli & Nuclear Envelope, Transformation
Order Info
Shipping Condition: Room Temperature
Compliance
Manufactured at facilities that comply with cGMP manufacturing requirements and that are certified to ISO 13485 and registered with FDA as medical device manufacturers.
Spécifications
Spécifications
| Concentration | 1 μg/mL |
| Form | Powder |
| Product Type | Antibiotic |
| Quantity | 5 mg |
For Research Use Only. Not for use in diagnostic procedures.