Research Products Corporation Acrylamide/bis-Acrylamide, 19:1 Ratio Solution, 500 Milliliters
Numéro de catalogue. 50488439
A 40% solution containing 38% (w/v) Acrylamide and 2% (w/v) bis-Acrylamide for a monomer to crosslinker ratio of 19:1.
WARNING! This product can expose you to Acrylamide, a chemical which is known to the State of California to cause cancer, developmental and reproductive harm. For more information go to www.P65Warnings.ca.gov.
Numéro de catalogue.
50488439
Fournisseur
Research Products CorporationCode fournisseur
A11400500.0
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
A 40% solution containing 38% (w/v) Acrylamide and 2% (w/v) bis-Acrylamide for a monomer to crosslinker ratio of 19:1.
WARNING! This product can expose you to Acrylamide, a chemical which is known to the State of California to cause cancer, developmental and reproductive harm. For more information go to www.P65Warnings.ca.gov.