missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide, >99%, Molecular biology reagent grade, MP Biomedicals™

Catalog No. ICN19398290
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19398290 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19398290 Supplier MP Biomedicals Inc Supplier No. 0219398290

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Color White
Density 1.127g/cm3 at 25°C
Quantity 500 g
Formula Weight 71.1
Percent Purity >99%
Grade Molecular Biology Reagent
Physical Form Crystalline Powder
Chemical Name or Material Acrylamide