missing translation for 'onlineSavingsMsg'
Learn More

Acridine, 97%

Catalog No. AAL0165714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0165714 25 g
AAL0165706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0165714 Supplier Thermo Scientific Chemicals Supplier No. L0165714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 260-94-6 | C13H9N | 179.222 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 260-94-6
Molecular Formula C13H9N
Molecular Weight (g/mol) 179.222
MDL Number MFCD00005025
InChI Key DZBUGLKDJFMEHC-UHFFFAOYSA-N
Synonym 9-azaanthracene, 2,3-benzoquinoline, acrydine, 10-azaanthracene, akridin, dibenzo b,e pyridine, 2,3,5,6-dibenzopyridine, akridin czech, benzo b quinoline
PubChem CID 9215
ChEBI CHEBI:36420
IUPAC Name acridine
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=N2

Specifications

Melting Point 107°C to 111°C
Density 1.005
Boiling Point 346°C
Quantity 25 g
UN Number UN2713
Beilstein 120200
Merck Index 14,122
Formula Weight 179.22
Percent Purity ≥98%
Chemical Name or Material Acridine
Hazard Category H302-H315-H319-H335-H340
Hazard Statement GHS H Statement
H341-H302-H315-H319-H335
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRIDINE
EINECSNumber 205-971-6
RTECSNumber AR7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only