Learn More
Waters Limited ACQUITY UPLC BEH Phenyl VanGuard Pre-column, 130, 1.7 m, 2.1 mm X 5 mm, 3/pk
Numéro de catalogue. 50782106
Encompass
ACQUITY BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This sorbent, along with the XBridge BEH Phenyl, provide you with the most chemiscally stable phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape. The VanGuard pre-column connects directly to the inlet of the ACQUITY UPLC column which minimizes extra columns volumes and reduces leaking by eliminating additional connections.
Numéro de catalogue. 50782106 Fournisseur Waters Limited Code fournisseur 186003979
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
ACQUITY BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This sorbent, along with the XBridge BEH Phenyl, provide you with the most chemiscally stable phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape. The VanGuard pre-column connects directly to the inlet of the ACQUITY UPLC column which minimizes extra columns volumes and reduces leaking by eliminating additional connections.