missing translation for 'onlineSavingsMsg'
Learn More

Acid Red 87 90.0+%, TCI America™

Catalog No. T003725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T003725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T003725G Supplier TCI America Supplier No. T003725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H12Br4Na2O8
Molecular Weight (g/mol) 745.904
MDL Number MFCD00005040
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Synonym Eosine, Tetrabromofluorescein Sodium Salt
PubChem CID 91886399
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]

Specifications

Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 691.86
Percent Purity ≥90.0% (E)
Chemical Name or Material Acid Red 87
EINECSNumber (5)-1511&(5)-3256
RTECSNumber LM5850000
TSCA Yes