missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Acid Fuchsin, pure, high purity biological stain
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Absorbance | 700 (at lambda min.) |
| Absorption Ratio | 1 to 1.4 |
| Infrared Spectrum | Authentic |
| Lambda Max | 544 to 550nm (in pH 3 acidified water) |
| Packaging | Glass bottle |
| Color | Green |
| Solubility Information | Solubility in water: 143g/L. Other solubilities: slightly soluble in alcohol |
| Quantity | 500 g |
| Physical Form | Powder or Crystals |
| Formula Weight | 585.52 |
| Show More |
Safety and Handling
| EINECSNumber | 221-816-5 |
| RTECSNumber | DD4737000 |
| TSCA | TSCA |
RUO – Research Use Only