missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Acid Fast Bacillus Stain Kits

Numéro de catalogue. 361014346
Modifier l'affichage
Click to view available options
Product Type:
Acid Fast Bacillus, Blue Secondary Stain Kit
Acid Fast Bacillus, Green Secondary Stain Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Product Type
361014346 Acid Fast Bacillus, Green Secondary Stain Kit
361014347 Acid Fast Bacillus, Blue Secondary Stain Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 361014346 Fournisseur Epredia™ Code fournisseur 87015
Il en reste null

Produce optimal staining while reducing precipitate with the Epredia™ Acid Fast Bacillus.

Acid-fast bacilli are stained bright red against a pale blue or pale green background.

Green Secondary Stain Kit includes:

  • Fast Green Stain Solution, 250mL
  • Carbol Fuchsin Solution, 125mL
  • Differentiating Solution, 500mL

Blue Secondary Stain Kit includes:

  • Loeffler Methylene Blue Stain Solution
  • Carbol Fuchsin Solution, 125mL
  • Differentiating Solution, 500mL

Spécifications

Product Type Acid Fast Bacillus, Green Secondary Stain Kit