missing translation for 'onlineSavingsMsg'
Learn More

Acid Blue 9, TCI America™

Catalog No. B079025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B079025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B079025G Supplier TCI America Supplier No. B079025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3844-45-9
Molecular Formula C37H34N2Na2O9S3
Molecular Weight (g/mol) 792.844
MDL Number MFCD00012141
InChI Key SGHZXLIDFTYFHQ-UHFFFAOYSA-L
Synonym Brilliant Blue FCF, Erioglaucin A
PubChem CID 19700
ChEBI CHEBI:82411
IUPAC Name disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Purple/Red
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 792.84
Chemical Name or Material Acid Blue 9
EINECSNumber (5)-1632&(5)-1732
RTECSNumber BQ4725000
TSCA Yes