missing translation for 'onlineSavingsMsg'
Learn More

Acid Black 1 97.0+%, TCI America™

Catalog No. A20975G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A20975G Supplier TCI America Supplier No. A20975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity acid black powder designed for electrophoresis applications.

Chemical Identifiers

CAS 1064-48-8
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.487
MDL Number MFCD00004017
InChI Key HKBVRFLHNUEVRO-UHFFFAOYSA-L
Synonym Amido Black 10B, Aminoschwarz 10B, Buffalo Black NBR, Naphthol Blue Black
PubChem CID 44134531
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 5 g
Physical Form Crystal/Powder
Formula Weight 616.49
Percent Purity ≥97.0% (HPLC)
Chemical Name or Material Acid Black 1 [for Electrophoresis]
EINECSNumber (5)-1875&(5)-5232
RTECSNumber QJ6196000
TSCA Yes