missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylsalicylic Acid 98.0+%, TCI America™
Supplier: TCI America A226225G
Specifications
| Acetylsalicylic Acid | |
| 134°C | |
| C9H8O4 | |
| 25 g | |
| aspirin, acetylsalicylic acid, 2-acetoxybenzoic acid, 2-acetyloxy benzoic acid, acetylsalicylate, o-acetylsalicylic acid, o-acetoxybenzoic acid, acylpyrin, polopiryna, easprin | |
| CC(=O)OC1=CC=CC=C1C(O)=O | |
| 180.16 | |
| CHEBI:15365 | |
| ≥98.0% (GC,T) |
| 50-78-2 | |
| White-Yellow | |
| MFCD00002430 | |
| 2811 | |
| BSYNRYMUTXBXSQ-UHFFFAOYSA-N | |
| 2-(acetyloxy)benzoic acid | |
| 2244 | |
| 180.16 | |
| Crystalline Powder |
Chemical Identifiers
| 50-78-2 | |
| 180.16 | |
| BSYNRYMUTXBXSQ-UHFFFAOYSA-N | |
| 2244 | |
| 2-(acetyloxy)benzoic acid |
| C9H8O4 | |
| MFCD00002430 | |
| aspirin, acetylsalicylic acid, 2-acetoxybenzoic acid, 2-acetyloxy benzoic acid, acetylsalicylate, o-acetylsalicylic acid, o-acetoxybenzoic acid, acylpyrin, polopiryna, easprin | |
| CHEBI:15365 | |
| CC(=O)OC1=CC=CC=C1C(O)=O |
Safety and Handling
EINECSNumber : (3)-1652
RTECSNumber : VO0700000
TSCA : Yes