missing translation for 'onlineSavingsMsg'
Learn More

Acetylsalicylic Acid 98.0+%, TCI America™

Catalog No. p-7117811
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A226225G 25 g
A2262500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A226225G Supplier TCI America Supplier No. A226225G
Only null left

Chemical Identifiers

CAS 50-78-2
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00002430
InChI Key BSYNRYMUTXBXSQ-UHFFFAOYSA-N
Synonym aspirin, acetylsalicylic acid, 2-acetoxybenzoic acid, 2-acetyloxy benzoic acid, acetylsalicylate, o-acetylsalicylic acid, o-acetoxybenzoic acid, acylpyrin, polopiryna, easprin
PubChem CID 2244
ChEBI CHEBI:15365
IUPAC Name 2-(acetyloxy)benzoic acid
SMILES CC(=O)OC1=CC=CC=C1C(O)=O

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 180.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Acetylsalicylic Acid
EINECSNumber (3)-1652
RTECSNumber VO0700000
TSCA Yes