missing translation for 'onlineSavingsMsg'
Learn More

Acetoxime 98.0+%, TCI America™

Catalog No. A0065500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0065500G 500 g
A006525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0065500G Supplier TCI America Supplier No. A0065500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 127-06-0
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00002118
InChI Key PXAJQJMDEXJWFB-UHFFFAOYSA-N
Synonym acetone oxime, acetoxime, propan-2-one oxime, 2-propanone, oxime, 2-propanone oxime, acetonoxime, acetone, oxime, beta-isonitrosopropane, acetoneoxime, ccris 5
PubChem CID 67180
ChEBI CHEBI:15349
IUPAC Name N-(propan-2-ylidene)hydroxylamine
SMILES CC(C)=NO

Specifications

Melting Point 62°C
Color White-Yellow
Boiling Point 133°C
Quantity 500 g
Formula Weight 73.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Acetoxime
EINECSNumber (2)-2479
RTECSNumber AL6825000
TSCA Yes